siRNA Details
| Gene Symbol: FLJ44715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126671.1 | - | 1 | 35 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI833237.1 | - | 1 | 70 | Chr. 10 - 73772157 - 73772897 [-] on Build GRCh38 | |

FLJ44715
Genomic Map