siRNA Details
| Gene Symbol: FHL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC011015.1 | - | 1 | 834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FHL5| Gene Symbol: FHL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC011015.1 | - | 1 | 834 | ||