siRNA Details
| Gene Symbol: ZNF181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026739.1 | XP_016882228.1 | 1 | 37 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF181| Gene Symbol: ZNF181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026739.1 | XP_016882228.1 | 1 | 37 | ||