siRNA Details
| Gene Symbol: CEBPB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125739.1 | 7 | 1786 | |||
| GenBank mRNA | AK129842.1 | - | 1 | 561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEBPB-AS1| Gene Symbol: CEBPB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125739.1 | 7 | 1786 | |||
| GenBank mRNA | AK129842.1 | - | 1 | 561 | ||