siRNA Details
| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129844.1 | - | 1 | 1343 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR744403.1 | - | 1 | 303 | Chr. 6 - 118561428 - 118561901 [+] on Build GRCh38 | |

PLN
Genomic Map