siRNA Details
| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002667.4 | NP_002658.1 | 2 | 2627 | ||
| GenBank mRNA | AK129844.1 | - | 1 | 934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLN| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002667.4 | NP_002658.1 | 2 | 2627 | ||
| GenBank mRNA | AK129844.1 | - | 1 | 934 | ||