siRNA Details
| Gene Symbol: SCAMP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105014.1 | 2 | 1392 | |||
| GenBank mRNA | AF086472.1 | - | 1 | 554 | ||
| AK129849.1 | - | 1 | 1471 | |||
| BC039455.1 | - | 2 | 1197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCAMP1-AS1| Gene Symbol: SCAMP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105014.1 | 2 | 1392 | |||
| GenBank mRNA | AF086472.1 | - | 1 | 554 | ||
| AK129849.1 | - | 1 | 1471 | |||
| BC039455.1 | - | 2 | 1197 | |||