siRNA Details
| Gene Symbol: DAOA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040247.1 | 2, 3 | 212 | |||
| GenBank mRNA | AY138548.1 | - | 2, 3 | 212 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DAOA-AS1| Gene Symbol: DAOA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040247.1 | 2, 3 | 212 | |||
| GenBank mRNA | AY138548.1 | - | 2, 3 | 212 | ||