siRNA Details
| Gene Symbol: LYRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017011084.1 | XP_016866573.1 | 4 | 3321 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK126470.1 | - | 2 | 3165 | Chr. 6 - 5031756 - 5054423 [-] on Build GRCh38 | |

LYRM4
Genomic Map