siRNA Details
Gene Symbol: MIR124-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109792.1 | 3 | 5443 | |||
GenBank mRNA | AK124231.1 | - | 1 | 1191 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIR124-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109792.1 | 3 | 5443 | |||
GenBank mRNA | AK124231.1 | - | 1 | 1191 |