siRNA Details
| Gene Symbol: MIR124-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109792.1 | 3 | 5584 | |||
| GenBank mRNA | AK124231.1 | - | 1 | 1332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR124-2HG| Gene Symbol: MIR124-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109792.1 | 3 | 5584 | |||
| GenBank mRNA | AK124231.1 | - | 1 | 1332 | ||