siRNA Details
| Gene Symbol: KANSL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005246334.4 | XP_005246391.1 | 6 | 2773 | ||
| XM_006712320.3 | XP_006712383.1 | 6 | 2855 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KANSL1L| Gene Symbol: KANSL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005246334.4 | XP_005246391.1 | 6 | 2773 | ||
| XM_006712320.3 | XP_006712383.1 | 6 | 2855 | |||