siRNA Details
| Gene Symbol: LOC101928150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120634.1 | 6 | 2422 | |||
| GenBank mRNA | CR749724.1 | - | 6 | 2427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928150| Gene Symbol: LOC101928150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120634.1 | 6 | 2422 | |||
| GenBank mRNA | CR749724.1 | - | 6 | 2427 | ||