siRNA Details
| Gene Symbol: EIF3J-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034170.1 | 1 | 573 | |||
| GenBank mRNA | AK092547.1 | - | 1 | 324 | ||
| BC026043.1 | - | 1 | 561 | |||
| BC042851.1 | - | 1 | 327 | |||
| BC066970.1 | - | 1 | 573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF3J-AS1| Gene Symbol: EIF3J-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034170.1 | 1 | 573 | |||
| GenBank mRNA | AK092547.1 | - | 1 | 324 | ||
| BC026043.1 | - | 1 | 561 | |||
| BC042851.1 | - | 1 | 327 | |||
| BC066970.1 | - | 1 | 573 | |||