siRNA Details
| Gene Symbol: CDK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC001274.1 | - | 9 | 1805 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK024379.1 | - | 1 | 3074 | Chr. 7 - 40066704 - 40070406 [+] on Build GRCh38 | |

CDK13
Genomic Map