siRNA Details
| Gene Symbol: KTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021278.1 | XP_016876767.1 | 1 | 737 | ||
| XM_017021281.1 | XP_016876770.1 | 1 | 737 | |||
| GenBank mRNA | BC041918.1 | - | 1 | 1014 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KTN1| Gene Symbol: KTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021278.1 | XP_016876767.1 | 1 | 737 | ||
| XM_017021281.1 | XP_016876770.1 | 1 | 737 | |||
| GenBank mRNA | BC041918.1 | - | 1 | 1014 | ||