siRNA Details
| Gene Symbol: ZDHHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008222.2 | NP_001008223.1 | 10 | 4376 | ||
| NM_016032.3 | NP_057116.2 | 11 | 4356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZDHHC9| Gene Symbol: ZDHHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008222.2 | NP_001008223.1 | 10 | 4376 | ||
| NM_016032.3 | NP_057116.2 | 11 | 4356 | |||