siRNA Details
Gene Symbol: ZDHHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008222.2 | NP_001008223.1 | 10 | 3946 | ||
NM_016032.3 | NP_057116.2 | 11 | 3926 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZDHHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008222.2 | NP_001008223.1 | 10 | 3946 | ||
NM_016032.3 | NP_057116.2 | 11 | 3926 |