siRNA Details
| Gene Symbol: LOC105375556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133929.1 | 2 | 647 | |||
| NR_133930.1 | 2 | 290 | ||||
| GenBank mRNA | BF188822.1 | - | 2 | 632 | ||
| BX385985.2 | - | 2 | 290 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375556| Gene Symbol: LOC105375556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133929.1 | 2 | 647 | |||
| NR_133930.1 | 2 | 290 | ||||
| GenBank mRNA | BF188822.1 | - | 2 | 632 | ||
| BX385985.2 | - | 2 | 290 | |||