siRNA Details
Gene Symbol: LOC101929057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111915.1 | 4 | 2977 | |||
GenBank mRNA | CR936797.1 | - | 4 | 2977 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111915.1 | 4 | 2977 | |||
GenBank mRNA | CR936797.1 | - | 4 | 2977 |