siRNA Details
| Gene Symbol: ANKRD44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195144.1 | NP_001182073.1 | 28 | 5671 | ||
| XM_005246945.1 | XP_005247002.1 | 28 | 6993 | |||
| XM_005246946.1 | XP_005247003.1 | 28 | 6939 | |||
| XM_006712838.3 | XP_006712901.1 | 28 | 7162 | |||
| XM_011512148.2 | XP_011510450.1 | 28 | 8874 | |||
| XM_017005252.1 | XP_016860741.1 | 28 | 8820 | |||
| GenBank mRNA | AK021632.1 | - | 1 | 1508 | ||

ANKRD44
Genomic Map