siRNA Details
| Gene Symbol: LOC101928650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096329.1 | - | 3 | 638 | ||
| BC065239.1 | - | 2 | 738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928650| Gene Symbol: LOC101928650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096329.1 | - | 3 | 638 | ||
| BC065239.1 | - | 2 | 738 | |||