siRNA Details
Gene Symbol: LOC101928314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125894.1 | 3 | 878 | |||
GenBank mRNA | BC043512.1 | - | 3 | 878 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125894.1 | 3 | 878 | |||
GenBank mRNA | BC043512.1 | - | 3 | 878 |