siRNA Details
| Gene Symbol: GNA14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121184.1 | 5 | 629 | |||
| GenBank mRNA | BC043514.2 | - | 4 | 595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNA14-AS1| Gene Symbol: GNA14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121184.1 | 5 | 629 | |||
| GenBank mRNA | BC043514.2 | - | 4 | 595 | ||