siRNA Details
| Gene Symbol: ZRANB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123114.1 | - | 1 | 368 | ||
| CR749325.1 | - | 1 | 3442 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZRANB2| Gene Symbol: ZRANB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123114.1 | - | 1 | 368 | ||
| CR749325.1 | - | 1 | 3442 | |||