siRNA Details
| Gene Symbol: LOC101928460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110758.1 | 3 | 1942 | |||
| GenBank mRNA | BC040893.1 | - | 3 | 1942 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417651.1 | - | 3 | 1942 | Chr. 1 - 29329620 - 29350114 [-] on Build GRCh38 | |

LOC101928460
Genomic Map