siRNA Details
| Gene Symbol: LOC101928460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110758.1 | 3 | 828 | |||
| GenBank mRNA | BC040893.1 | - | 3 | 828 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM554569.1 | - | 3 | 853 | Chr. 1 - 29330735 - 29350114 [-] on Build GRCh38 | |

LOC101928460
Genomic Map