siRNA Details
| Gene Symbol: TNRC6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006721996.3 | XP_006722059.3 | 1 | 3294 | ||
| XM_006721997.3 | XP_006722060.3 | 1 | 3294 | |||
| XM_006722000.3 | XP_006722063.3 | 1 | 3294 | |||
| XM_017024906.1 | XP_016880395.1 | 1 | 3294 | |||
| XM_017024907.1 | XP_016880396.1 | 1 | 3294 | |||
| XM_017024908.1 | XP_016880397.1 | 1 | 3294 | |||
| GenBank mRNA | BC035105.1 | - | 1 | 2188 | ||
| BC051468.1 | - | 1 | 2183 | |||

TNRC6C
Genomic Map