siRNA Details
| Gene Symbol: LOC645645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126090.1 | - | 1 | 1342 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC645645| Gene Symbol: LOC645645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126090.1 | - | 1 | 1342 | ||