siRNA Details
| Gene Symbol: RGL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126263.1 | - | 1 | 803 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF351589.1 | - | 1 | 130 | Chr. 19 - 11395277 - 11395453 [-] on Build GRCh38 | |

RGL3
Genomic Map