siRNA Details
| Gene Symbol: LOC257396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034107.2 | 2 | 1060 | |||
| GenBank mRNA | AK097893.1 | - | 2 | 1045 | ||
| BC041894.1 | - | 2 | 1003 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC257396| Gene Symbol: LOC257396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034107.2 | 2 | 1060 | |||
| GenBank mRNA | AK097893.1 | - | 2 | 1045 | ||
| BC041894.1 | - | 2 | 1003 | |||