siRNA Details
| Gene Symbol: LOC257396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034107.2 | 3 | 1971 | |||
| NR_104654.1 | 2 | 1176 | ||||
| GenBank mRNA | AK097893.1 | - | 3 | 1956 | ||
| BC041894.1 | - | 2 | 2738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC257396| Gene Symbol: LOC257396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034107.2 | 3 | 1971 | |||
| NR_104654.1 | 2 | 1176 | ||||
| GenBank mRNA | AK097893.1 | - | 3 | 1956 | ||
| BC041894.1 | - | 2 | 2738 | |||