siRNA Details
| Gene Symbol: LOC257396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034107.2 | 3 | 1376 | |||
| NR_104654.1 | 2 | 581 | ||||
| GenBank mRNA | AK097893.1 | - | 3 | 1361 | ||
| BC041894.1 | - | 2 | 2143 | |||
| BQ637503.1 | - | 2 | 546 | |||
| DR158823.1 | - | 2 | 536 | |||

LOC257396
Genomic Map