siRNA Details
| Gene Symbol: CLDN10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046533.1 | 4 | 397 | |||
| GenBank mRNA | BC024972.1 | - | 4 | 398 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG574725.1 | - | 4 | 395 | Chr. 13 - 95479522 - 95533902 [-] on Build GRCh38 | |

CLDN10-AS1
Genomic Map