siRNA Details
| Gene Symbol: BOD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL137652.1 | - | 1 | 454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BOD1L1| Gene Symbol: BOD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL137652.1 | - | 1 | 454 | ||