siRNA Details
| Gene Symbol: DAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092836.1 | - | 1 | 650 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK123803.1 | - | 1 | 1489 | Chr. 5 - 39372644 - 39375053 [-] on Build GRCh38 | |

DAB2
Genomic Map