siRNA Details
| Gene Symbol: LINC01527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042056.1 | - | 2 | 1681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01527| Gene Symbol: LINC01527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042056.1 | - | 2 | 1681 | ||