siRNA Details
Gene Symbol: LOC105376204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015409.1 | XP_016870898.1 | 1 | 3928 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK023447.1 | - | 1 | 1560 | Chr. 9 - 106858756 - 106860647 [-] on Build GRCh38 |
