siRNA Details
| Gene Symbol: LOC105376204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017015409.1 | XP_016870898.1 | 1 | 2749 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK023447.1 | - | 1 | 381 | Chr. 9 - 106858756 - 106860647 [-] on Build GRCh38 | |

LOC105376204
Genomic Map