siRNA Details
| Gene Symbol: WBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038789.1 | - | 3 | 364 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI461473.1 | - | 3 | 369 | Chr. 22 - 41998793 - 42002910 [+] on Build GRCh38 | |

WBP2NL
Genomic Map