siRNA Details
| Gene Symbol: LOC101927701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110271.1 | 1 | 43 | |||
| GenBank mRNA | BC028245.1 | - | 1 | 54 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI861426.1 | - | 1 | 42 | Chr. 2 - 67796054 - 67825561 [-] on Build GRCh38 | |

LOC101927701
Genomic Map