siRNA Details
| Gene Symbol: LOC101927701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110271.1 | 3 | 677 | |||
| GenBank mRNA | BC028245.1 | - | 3 | 688 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927701| Gene Symbol: LOC101927701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110271.1 | 3 | 677 | |||
| GenBank mRNA | BC028245.1 | - | 3 | 688 | ||