siRNA Details
| Gene Symbol: LINC01514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120620.1 | 6 | 926 | |||
| GenBank mRNA | BC038784.1 | - | 6 | 925 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000434189.2 | - | 6 | 925 | Chr. 10 - 101176323 - 101194147 [+] on Build GRCh38 | |

LINC01514
Genomic Map