siRNA Details
Gene Symbol: LINC01514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120620.1 | 4, 5 | 694 | |||
GenBank mRNA | BC038784.1 | - | 4, 5 | 693 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120620.1 | 4, 5 | 694 | |||
GenBank mRNA | BC038784.1 | - | 4, 5 | 693 |