siRNA Details
| Gene Symbol: LINC01514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120620.1 | 2 | 400 | |||
| GenBank mRNA | BC038784.1 | - | 2 | 399 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI464847.1 | - | 2 | 402 | Chr. 10 - 101176321 - 101191431 [+] on Build GRCh38 | |

LINC01514
Genomic Map