siRNA Details
| Gene Symbol: LOC105378828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135837.1 | 1 | 161 | |||
| NR_135838.1 | 1 | 161 | ||||
| GenBank mRNA | BC034638.1 | - | 1 | 130 | ||
| HY051678.1 | - | 1 | 165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378828| Gene Symbol: LOC105378828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135837.1 | 1 | 161 | |||
| NR_135838.1 | 1 | 161 | ||||
| GenBank mRNA | BC034638.1 | - | 1 | 130 | ||
| HY051678.1 | - | 1 | 165 | |||