siRNA Details
| Gene Symbol: LOC105378828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135838.1 | 1 | 892 | |||
| GenBank mRNA | BC034638.1 | - | 1 | 861 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI913017.1 | - | 1 | 418 | Chr. 1 - 86703298 - 86704018 [-] on Build GRCh38 | |

LOC105378828
Genomic Map