siRNA Details
| Gene Symbol: LOC219690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055023.1 | - | 3 | 726 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC219690| Gene Symbol: LOC219690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055023.1 | - | 3 | 726 | ||