siRNA Details
| Gene Symbol: SLC47A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135624.1 | 1 | 625 | |||
| XM_017024222.1 | XP_016879711.1 | 1 | 488 | |||
| GenBank mRNA | AK123589.1 | - | 1 | 484 | ||
| BX648861.1 | - | 1 | 625 | |||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: SLC47A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135624.1 | 1 | 625 | |||
| XM_017024222.1 | XP_016879711.1 | 1 | 488 | |||
| GenBank mRNA | AK123589.1 | - | 1 | 484 | ||
| BX648861.1 | - | 1 | 625 | |||
SLC47A2
Genomic Map