siRNA Details
| Gene Symbol: TMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021741.1 | - | 1305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMF1| Gene Symbol: TMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021741.1 | - | 1305 | |||